Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535039
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535039 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535039 Supplier Invitrogen™ Supplier No. PA535039
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EAP2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

This gene encodes a member of a superfamily of divalent cation-dependent phosphodiesterases. The encoded protein associates with CD40, tumor necrosis factor (TNF) receptor-75 and TNF receptor associated factors (TRAFs), and inhibits nuclear factor-kappa-B activation. This protein has sequence and structural similarities with APE1 endonuclease, which is involved in both DNA repair and the activation of transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EAP2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TDP2
Gene Accession No. O95551
Gene Alias 5'-Tyr-DNA phosphodiesterase; 5'-tyrosyl-DNA phosphodiesterase; AD022; AD-022; D13Ertd656e; dJ30M3.3; EAP2; EAPII; ETS1-associated protein 2; ETS1-associated protein II; hTDP2; MGC111021; MGC9099; RP1-30M3.3; Tdp2; TRAF and TNF receptor associated protein; TRAF and TNF receptor-associated protein; TTRAP; Tyr-DNA phosphodiesterase 2; tyrosyl-DNA phosphodiesterase 2; Tyrosyl-RNA phosphodiesterase; VPg unlinkase
Gene Symbols TDP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 246-272 amino acids from the C-terminal region of human EAPII.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51567
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.