Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588177
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588177 Supplier Invitrogen™ Supplier No. PA588177
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MELGSCLEGG REAAEEEGEP EVKKRRLLCV EFASVASCDA AVAQCFLAEN DWEMERALNS YFEPPVEESA LERRPETISE PKTYVDLTNE ETTDSTTSKI SPSEDTQQEN GSMFSLITWN IDGLDLNNLS ERARGVCSYL ALYSPDVIFL QEVIPPYYSY LKKRSSNYEI ITGHEEGYFT AIMLKKSRVK LKSQEIIPFP; Positive Samples: HepG2, HeLa, Mouse brain; Cellular Location: Cytoplasm, Nucleus, PML body, nucleolus.

This gene encodes a member of a superfamily of divalent cation-dependent phosphodiesterases. The encoded protein associates with CD40, tumor necrosis factor (TNF) receptor-75 and TNF receptor associated factors (TRAFs), and inhibits nuclear factor-kappa-B activation. This protein has sequence and structural similarities with APE1 endonuclease, which is involved in both DNA repair and the activation of transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EAP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TDP2
Gene Accession No. O95551, Q9JJX7
Gene Alias 5'-Tyr-DNA phosphodiesterase; 5'-tyrosyl-DNA phosphodiesterase; AD022; AD-022; D13Ertd656e; dJ30M3.3; EAP2; EAPII; ETS1-associated protein 2; ETS1-associated protein II; hTDP2; MGC111021; MGC9099; RP1-30M3.3; Tdp2; TRAF and TNF receptor associated protein; TRAF and TNF receptor-associated protein; TTRAP; Tyr-DNA phosphodiesterase 2; tyrosyl-DNA phosphodiesterase 2; Tyrosyl-RNA phosphodiesterase; VPg unlinkase
Gene Symbols TDP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human TDP2 (NP_0576982).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51567, 56196
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.