Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EAP30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531609
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531609 Supplier Invitrogen™ Supplier No. PA531609
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (98%), Rat (99%), Zebrafish (82%), Xenopus laevis (84%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ELL encodes an RNA polymerase II transcription factor that undergoes frequent translocation in acute myeloid leukemia (AML). In addition to its elongation activity, ELL contains a novel type of RNA polymerase II interaction domain that is capable of repressing polymerase activity in promoter-specific transcription. EAP30 is a subunit of the ELL complex. EAP30 can interact with ELL and derepress ELL's inhibitory activity in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EAP30
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SNF8
Gene Accession No. Q96H20
Gene Alias D11Moh34; Dot3; Eap30; EAP30 subunit of ELL complex; ELL-associated protein of 30 kDa; ESCRT-II complex subunit VPS22; hVps22; RGD1310144; si:dkey-220f10.1; snf8; SNF8 subunit of ESCRT-II; SNF8, ESCRT-II complex subunit; SNF8, ESCRT-II complex subunit, homolog; SNF8, ESCRT-II complex subunit, homolog (S. cerevisiae); vacuolar sorting snf8 ELL associated of 30kDa; vacuolar-sorting protein SNF8; VPS22; zgc:101578
Gene Symbols SNF8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SNF8. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11267
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.