Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EAP30 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540389
Description
Peptide sequence: MHRRGVGAGA IAKKKLAEAK YKERGTVLAE DQLAQMSKQL DMFKTNLEEF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
ELL encodes an RNA polymerase II transcription factor that undergoes frequent translocation in acute myeloid leukemia (AML). In addition to its elongation activity, ELL contains a novel type of RNA polymerase II interaction domain that is capable of repressing polymerase activity in promoter-specific transcription. EAP30 is a subunit of the ELL complex. EAP30 can interact with ELL and derepress ELL's inhibitory activity in vitro.
Specifications
EAP30 | |
Polyclonal | |
Unconjugated | |
SNF8 | |
D11Moh34; Dot3; Eap30; EAP30 subunit of ELL complex; ELL-associated protein of 30 kDa; ESCRT-II complex subunit VPS22; hVps22; RGD1310144; si:dkey-220f10.1; snf8; SNF8 subunit of ESCRT-II; SNF8, ESCRT-II complex subunit; SNF8, ESCRT-II complex subunit, homolog; SNF8, ESCRT-II complex subunit, homolog (S. cerevisiae); vacuolar sorting snf8 ELL associated of 30kDa; vacuolar-sorting protein SNF8; VPS22; zgc:101578 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11267 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96H20 | |
SNF8 | |
Synthetic peptide directed towards the N-terminal of human EAP30 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction