Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EAPP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551774
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551774 Supplier Invitrogen™ Supplier No. PA551774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLLHGTPDQK RKLIRECLTG ESESSSEDEF EKEMEAELNS TMKTMEDKLS SLGTGSSSGN GKVATAPTRY YDDIYFDSDS EDEDRAVQVT KKKKKKQHKI PTNDELLYDP E Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

This protein may play an important role in the fine-tuning of both major E2F1 activities, the regulation of the cell-cycle and the induction of apoptosis. Promotes S-phase entry, and inhibits p14(ARP) expression. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. The C-terminal of EAPP binds the N-terminal of E2F1 and also interacts with E2F2 and E2F3, but not E2F4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EAPP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EAPP
Gene Accession No. Q56P03
Gene Alias 1810011O16Rik; BM036; BM-036; c14orf11; E2F associated phosphoprotein; E2F-associated phosphoprotein; Eapp; hypothetical protein LOC327318; RGD1309624; Unknown (protein for MGC:139672); wu:fd21g02; zgc:65820
Gene Symbols EAPP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EAPP. Recombinant protein control fragment (Product #RP-89046).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55837
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.