Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EAR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590335
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590335 Supplier Invitrogen™ Supplier No. PA590335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAMVTGGWGG PGGDTNGVDK AGGYPRAAED DSASPPGAAS DAEPGDEERP GLQVD; Positive Samples: Mouse liver, Rat liver; Cellular Location: Nucleus.

Transcription factor predominantly involved in transcriptional repression. Binds to promoter/enhancer response elements that contain the imperfect 5'-AGGTCA-3' direct or inverted repeats with various spacings which are also recognized by other nuclear hormone receptors. Involved in modulation of hormonal responses. Represses transcriptional activity of the lutropin-choriogonadotropic hormone receptor/LHCGR gene, the renin/REN gene and the oxytocin-neurophysin/OXT gene. Represses the triiodothyronine-dependent and -independent transcriptional activity of the thyroid hormone receptor gene in a cell type-specific manner. The corepressing function towards thyroid hormone receptor beta/THRB involves at least in part the inhibition of THRB binding to triiodothyronine response elements (TREs) by NR2F6. Inhibits NFATC transcription factor DNA binding and subsequently its transcriptional activity. Acts as transcriptional repressor of IL-17 expression in Th-17 differentiated CD4(+) T cells and may be involved in induction and/or maintenance of peripheral immunological tolerance and autoimmunity. Involved in development of forebrain circadian clock; is required early in the development of the locus coeruleus (LC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EAR2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NR2F6
Gene Accession No. O09017, P10588, P43136
Gene Alias AV090102; avian erythroblastic leukemia viral (v-erb-a) oncogene homolog-like 2; COUP transcription factor 3; COUPg; COUP-TF3; EAR2; EAR-2; Erba related 2; ERBAL2; ERBA-related gene-2; Nr2f6; Nuclear receptor subfamily 2 group F member 6; nuclear receptor subfamily 2, group F, member 6; nuclear receptor V-erbA-related; orphan nuclear receptor EAR-2; Ovalbumin upstream promoter gamma nuclear receptor; ovalbumin upstream promoter gamma nuclear receptor rCOUPg; v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 2; V-erbA-related protein 2; V-erbA-related protein EAR-2
Gene Symbols NR2F6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-55 of human NR2F6 (NP_0052252).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13864, 2063, 245980
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.