Learn More
EARS2 Mouse anti-Human, Clone: OTI2C1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810522

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Glutamyl-tRNA synthetase (GluRS or EARS2) a class I aminoacyl-tRNA synthetase (aaRS), is primarily responsible for the glutamylation of tRNAGlu. It is part of the ""minimal set"" of seventeen aaRSs found in every living organism and its presence is essential for the viability of cells.Specifications
EARS2 | |
Monoclonal | |
Unconjugated | |
EARS2 | |
COXPD12, MSE1, gluRS | |
Mouse | |
Affinity Chromatography | |
RUO | |
124454 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2C1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q5JPH6 | |
EARS2 | |
Full length human recombit protein of human EARS2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.