Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560312
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560312 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560312 Supplier Thermo Scientific Supplier No. PA560312
Add to Cart
Add to Cart

EARS2 Polyclonal Antibody

EARS2 Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen EARS2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ears2
Gene Accession No. Q5JPH6
Gene Alias 3230401I01Rik; COXPD12; Ears2; GluRS; glutamate tRNA ligase 2, mitochondrial; glutamate--tRNA ligase; Glutamyl-tRNA synthetase; glutamyl-tRNA synthetase 2 (mitochondrial)(putative); glutamyl-tRNA synthetase 2 mitochondrial (putative); glutamyl-tRNA synthetase 2, mitochondrial; Kiaa1970; mKIAA1970; MSE1; Probable glutamate--tRNA ligase, mitochondrial; probable glutamyl-tRNA synthetase, mitochondrial; RGD1307904
Gene Symbols Ears2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EARS2. Recombinant protein control fragment (Product #RP-98632).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124454
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.