Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113954
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113954 Supplier Invitrogen™ Supplier No. PA5113954
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TAKHLLLYQA LGWQPPHFAH LPLLLNRDGS KLSKRQGDVF LEHFAADGFL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 100%; Yeast: 91%; Zebrafish: 93%.

Glutamyl-tRNA synthetase (GluRS or EARS2) a class I aminoacyl-tRNA synthetase (aaRS), is primarily responsible for the glutamylation of tRNAGlu. It is part of the minimal set of seventeen aaRSs found in every living organism and its presence is essential for the viability of cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EARS2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ears2
Gene Accession No. Q5JPH6
Gene Alias 3230401I01Rik; COXPD12; Ears2; GluRS; glutamate tRNA ligase 2, mitochondrial; glutamate--tRNA ligase; Glutamyl-tRNA synthetase; glutamyl-tRNA synthetase 2 (mitochondrial)(putative); glutamyl-tRNA synthetase 2 mitochondrial (putative); glutamyl-tRNA synthetase 2, mitochondrial; Kiaa1970; mKIAA1970; MSE1; Probable glutamate--tRNA ligase, mitochondrial; probable glutamyl-tRNA synthetase, mitochondrial; RGD1307904
Gene Symbols Ears2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human EARS2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124454
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.