Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Easy Grip Disposable Polystyrene Sterile Bottle PROMO
Shop All Corning Products
Click to view available options
Capacity (Metric):
150 mL
250 mL
500 mL
1 L
Description
- Disposable polystyrene bottles for storage of media, buffers, and other aqueous solutions
- Corning low profile style has easy grip sides that facilitate handling
- Plug seal caps (45 mm) provide an airtight seal and help minimize the risk of contamination.
- Bottles can be used with Corning vacuum filtration systems with 45 mm neck sizes (Cat. Nos. 430626, 430627, 431161, 430049, 430513, 430514,431118, 430015, and 431174).
- Sterile
- Non-pyrogenic
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| For Use With (Application) | Storage of media, buffers and other aqueous solutions |
| Includes | Orange HDPE plug seal |
| Material | Polystyrene |
| No. per Case | 24 |
| No. per Pack | 2 |
| Sterility | Sterile |
| Disposable | Single-use |
| Mouth | Narrow |
| Product Type | Media Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction