Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EASY-Spray™ PepMap™ Neo UHPLC Columns

Catalog No. 03255023
Change view
Click to view available options
Length (Metric):
150 mm
500 mm
750 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
03-255-023 750 mm
03-255-021 150 mm
03-255-022 500 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-255-023 Supplier Thermo Scientific™ Supplier No. ES75750PN
Add to Cart
Add to Cart

Separate challenging peptide mapping samples with Thermo Scientific™ EASY-Spray™ PepMap™ Neo UHPLC columns.

Separate challenging peptide mapping samples using Thermo Scientific™ EASY-Spray™ PepMap™ Neo UHPLC columns. These columns feature easy connectivity, high reproducibilty, and excellent separations. Our Neo columns provide 1500 bar pressure capability, improved column-to-column consistency, and increased efficiency. The column media is manufactured and selected to exacting standards and packed at high pressure, resulting in enhanced peak symmetry, resolution, and column-to-column reproducibility that allows you to obtain greater sample coverage and sample insights. The Click-and-Spray format of EASY-Spray columns enables perfect connections to our mass spectrometers every time.

Benefits of PepMap Neo Columns

  • Brilliant Connectivity — EASY-Spray™ columns click into position with ease.
  • Brilliant Reproducibility — Precise manufacturing protocols for packing media improve column-to-column consistency for reproducible analysis every day.
  • Brilliant Separation — Ultra-high pressure packing increases column efficiency, allowing you to run at up to 1500 bar for enhanced peak symmetry and resolution, providing higher sample capacity for more sample insights.
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column, EASY-Spray™
Diameter (Metric) 75 μm
Length (Metric) 750 mm
Max. Pressure 22,000 psi (1500 bar)
Particle Size 2 μm
pH 2 to 8
Pore Size 100 Å
Surface Area 450 m2/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Product Line Easy Spray
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 15%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Description Easy-Spray™ PepMap™ Neo 2 μm C18 75 μm X 750 mm
For Use With (Application) Bottom-up Proteomics, LC-MS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.