Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Easy-Adapt™ CHO K1 CD Medium
GREENER_CHOICE

Catalog No. p-7084484
Click to view available options
Quantity:
10 L
20 L

Chemically defined (CD) medium

Rich in carbohydrates, amino acids, and vitamins, and specifically designed to provide the nutrients CHO cells need.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Protect from light.
Cell Line CHO, CHO-K1
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Powder
Product Type Cell Culture Media
Format BPC
Quantity 20 L
Source Animal origin-free
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.