Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EasyPep™ 96 MS Sample Prep Kit, 96 Reactions PROMO

Catalog No. PIA45733PM
Click to view available options
:
Thermo Scientific™ EasyPep™ 96 MS Sample Prep Kit

Generate high quality proteomic samples for MS analysis in less than four hours using optimized EasyPep MS Sample Prep kits.

  • Complete—includes pre-formulated reagents for lysis through digestion, peptide clean-up columns, and an optimized protocol for up to 20 samples
  • Optimized—streamlined protocol and reagents minimize the number of steps and time time needed to process samples
  • Flexible—reagents and protocol have been verified using cells, plasma, and tissue samples for 10 to 100 μg samples
  • Time-saving—sample processing time reduced from more than one day to less than three hours
  • Compatible—final preparation is ready for direct MS analysis and other downstream applications, including TMT labeling

The sample preparation of peptides for MS analysis is complex, with numerous steps and varied protocols, resulting in sample variability in both quality and quantity. This kit has been designed to improve reproducibility while saving hands-on and processing time. The number of steps and processing time have been reduced through the addition of Universal Nuclease to reduce viscosity from nucleic acids (replacing sonication), a rapid 'one pot' reduction/alkylation solution for cysteine modification (carbamidomethylation, + 57.02), and a trypsin/Lys-C protease mix for more complete digestion. In addition, the kit includes Peptide Clean-up columns and buffers to prepare detergent-free peptide samples for MS analysis. The optimized reagents and protocol produce high-quality peptides that are compatible with TMT labeling and other downstream applications, or are ready for MS analysis.

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Sample Preparation Kit
Product Line EasyPep™
Content And Storage
  • Lysis Solution, 25 mL, store at 4°C
  • Universal Nuclease, 25 kU, store at -20°C
  • Reduction Solution, 7 mL, store at 4°C
  • Alkylation Solution, 7 mL, store at 4°C
  • Enzyme Reconstitution solution, 7 mL, store at 4°C
  • Pierce Trypsin/Lys-C Protease Mix, MS Grade, 5 x 100 μg, store at -20°C
  • Digestion Stop Solution, 7 mL, store at 4°C
  • Peptide Clean-Up Plate, 1 Ea., store at 4°C
  • Wash and Elution Plate, 1 Ea., store at 4°C
  • Sample Prep Plate, 1 Ea., store at 4°C
  • Wash Solution A, 40 mL, store at 4°C
  • Wash Solution B, 3 x 27 mL, store at 4°C
  • Elution Solution, 2 x 20 mL, store at 4°C
Label or Dye Unlabeled
Shipping Condition Wet Ice
Starting Material Protein Samples
Final Product Type Peptides
Detection Method Mass Spectrometry
Format Kit
Workflow Step Protein Digestion
Quantity 96 Reactions
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.