Learn More
Thermo Scientific™ EasyPep™ Maxi Sample Prep Kit, 8 Reactions PROMO
Description
- Complete—includes pre-formulated reagents for lysis through digestion, peptide clean-up columns, and an optimized protocol for up to 20 samples
- Optimized—streamlined protocol and reagents minimize the number of steps and time time needed to process samples
- Flexible—reagents and protocol have been verified using cells, plasma, and tissue samples for 10 to 100 μg samples
- Time-saving—sample processing time reduced from more than one day to less than three hours
- Compatible—final preparation is ready for direct MS analysis and other downstream applications, including TMT labeling
The sample preparation of peptides for MS analysis is complex, with numerous steps and varied protocols, resulting in sample variability in both quality and quantity. This kit has been designed to improve reproducibility while saving hands-on and processing time. The number of steps and processing time have been reduced through the addition of Universal Nuclease to reduce viscosity from nucleic acids (replacing sonication), a rapid 'one pot' reduction/alkylation solution for cysteine modification (carbamidomethylation, + 57.02), and a trypsin/Lys-C protease mix for more complete digestion. In addition, the kit includes Peptide Clean-up columns and buffers to prepare detergent-free peptide samples for MS analysis. The optimized reagents and protocol produce high-quality peptides that are compatible with TMT labeling and other downstream applications, or are ready for MS analysis.
Specifications
Specifications
For Use With (Equipment) | Mass Spectrometer |
Product Type | Sample Preparation Kit |
Product Line | EasyPep™ |
Content And Storage | Lysis Solution, 25 mL, Store at 4°C, Universal Nuclease, 25 kU, Store at -20°C, Reduction Solution, 7 mL, Store at 4°C, Alkylation Solution, 7 mL, Store at 4°C, Enzyme Reconstititution Solution, 7 mL, Store at 4°C, Pierce™ Trypsin/Lys-C Protease Mix, MS Grade, 8 x 100 μg, Store at -20°C, Digestion Stop Solution, 7 mL, Store at 4°C, Peptide Clean-Up Columns, 8 Ea. Store at 4°C, Wash Solution A, 40 mL, Store at 4°C, Wash Solution B, 3 x 27 mL, Store at 4°C, Elution Solution, 2 x 20 mL, Store at 4°C, Low Protein Binding Collection Tubes, 2 mL, 40 Ea. Store at 4°C |
Label or Dye | Unlabeled |
Shipping Condition | Wet Ice |
Starting Material | Protein Samples |
Final Product Type | Peptides |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Digestion |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.