Learn More
Interscience EasySpiral
SDP
The easySpiral plater uses the Spiral technique for plating Petri dishes.
$26700.00 - $45850.00
Specifications
Type Automatic Plater
View More Specs
Products 4
Catalog Number Mfr. No. Product Line Price Quantity & Availability  
Catalog Number Mfr. No. Product Line Price Quantity & Availability  
16-949-030
SDP
View Documents
Interscience
412000
EasySpiral
Each for $26,700.00
Only null left
Add to Cart
16-949-051
SDP
View Documents
Interscience
413000
EasySpiral Pro
Each for $39,700.00
Only null left
Add to Cart
16-948-919
SDP
View Documents
Interscience
413090
EasySpiral Pro Milk
Each for $42,800.00
Only null left
Add to Cart
16-948-888
SDP
View Documents
Interscience
414000
EasySpiral Dilute
Each for $45,850.00
Only null left
Edge
Add to Cart
Description
Description
EasySpiral is an automatic plater of Petri dishes. It allows plating of a sample with 300 to 1.3x10^5 CFU/mL on one Petri dish, without prior dilution. Plated volumes are of 50 and 100 μL.
  • Syringe capacity: 1000 μL
  • Plating modes: exponential, circle
  • Counting range: from 300 to 1.3x10^5 CFU/mL
  • Plating cycle time: 25 seconds
EasySpiral Pro is an automatic plater of Petri dishes. It allows plating of a sample of 100 to 1x10^7 CFU/mL on one Petri dish, without prior dilution. Traceability is ensured and volumes are programmable via USB connection.
  • Traceability and programmable volumes
  • Plating modes: exponential, constant, circle
  • Counting range: from 100 to 1x10^7 CFU/mL
  • Plating cycle time: 25 seconds
EasySpiral Pro Milk is an automatic Petri dish plater. It allows plating of a sample of 100 to 1x10^7 CFU/mL on a single Petri dish, without prior dilution. EasySpiral Pro Milk is particularly suitable for the dairy industry in analyzing raw materials and payment for milk.
  • Sample intake in the sample holder
  • Plating modes: exponential, pour, circle
  • Counting range: from 100 to 1x10^7 CFU mL
  • Plating cycle time: 25 seconds
EasySpiral Dilute is an automatic diluter and plater of Petri dishes. It allows 1/10th serial dilutions and plating up to 6 log. dilution on 1 single Petri dish. For a sample of 30 to 1 x 10^12 bacteria, no more manual dilutions and plating is automatic.
  • Automatic cycle of 5 successive dilutions at 1/10th
  • Plating modes: exponential, constant, circle, pour plate
  • Count range: from 30 to 1x10^12 CFU/mL
  • Plating cycle time: 134 seconds
Specifications
Specifications
Automatic Plater
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.