Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interscience EasySpiral PTFE Filters
SDP

Catalog No. 16948984
Click to view available options
:
EasySpiral PTFE Filters

PTFE filters for use with EasySpiral platers.

  • PTFE filters for EasySpiral
  • Set of 5
  • Autoclavable
  • 0.2 μm
  • For use with GL45 bottle

Specifications

For Use With (Equipment) GL45 Bottle
Material (Membrane) PTFE
Pore Size 0.2 μm
Sterility Non-sterile
No. per Pack 5/Pk.
Type Filter
Diameter (Metric) 25 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.