Learn More
Thermo Scientific™ EasyStart™ PCR Mix-in-a-Tube
Description
Reduce tedious PCR set-up protocols to a single step using Thermo Scientific™ EasyStart™ PCR Mix-in-a-tube.
Features:
- Delivers better specificity and yield than conventionally prepared mixtures
- Compatible with a wide range of enzymes
- Wax layer provides safe, easy storage of reagent mixture, prevents oxidation and evaporation, and eliminates wasted reagents
Ordering Information:
Use 0.2mL tubes for 0.2mL heating blocks and 0.5mL tubes for 0.5 or 0.6mL heating blocks.
Specifications
Specifications
| Description | EasyStart Micro 100 |
| Volume (Metric) | 0.2 mL |
| Storage Requirements | Tubes can be stored at ambient temperatures for extended periods; To remove PCR product, simply pierce the wax layer with a pipet tip |
| Reaction Volume | 50 to 100 μL |
| Product Type | Tube |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.