Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EB1 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. FEPA5145291
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145291 200 μL
FEPA5145292 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145291 Supplier Invitrogen™ Supplier No. PA5145291
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EB1 family proteins are evolutionarily conserved proteins that bind microtubule plus-ends and centrosomes and regulate the dynamics and organization of microtubules. The MAPRE genes encode the EB1 family proteins. The human MAPRE genes MAPRE1, MAPRE2, and MAPRE3 code for EB1, RP1, and EBF3 proteins, respectively. EB1 family members appear to be expressed ubiquitously. The EB1 (MAPRE1) gene, at 20q11. 2, is fused to MLL in an adult patient with pro-B acute lymphoblastic leukemia. EB1 is a microtubule-associated protein that interacts with the colorectal adenomatous polyposis coli tumor-suppressor protein and plays important roles in regulating microtubule dynamics, cell polarity, and chromosome stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EB1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene Mapre1
Gene Accession No. Q15691
Gene Alias 5530600P05Rik; adenomatosis polyposis coli binding protein Eb1; adenomatous polyposis coli-binding protein EB1; AI462499; AI504412; APC-binding protein EB1; AW260097; BIM1p; D2Ertd459e; Eb1; end-binding protein 1; Mapre1; MGC117374; MGC129946; microtubule associated protein RP/EB family member 1; Microtubule-associated protein RP/EB family member 1; microtubule-associated protein RP/EB family member 1-like protein; microtubule-associated protein, RP/EB family, member 1
Gene Symbols Mapre1
Host Species Rabbit
Immunogen Recombinant Human MAPRE1 protein, Ala2-Tyr268 (Accession #Q15691).
Purification Method Protein A, Antigen Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22919, 733687
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.