Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EB2 Antibody (OTI1F3), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271650MFV450
Description
EB2 Monoclonal antibody specifically detects EB2 in Human samples. It is validated for Flow CytometrySpecifications
EB2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
OTI1F3 | |
Flow Cytometry | |
APC-binding protein EB2, EB1, EB2APC-binding protein EB1, End-binding protein 2, microtubule-associated protein, RP/EB family, member 2, RP1microtubule-associated protein RP/EB family member 2, T-cell activation protein, EB1 family | |
Full length human recombinant protein of human MAPRE2 (NP_055083) produced in HEK293T cell. | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
10982 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction