Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EB2 (MAPRE2) Monoclonal Antibody (OTI5F3), TrueMAB™, OriGene
SDP

Catalog No. 502541198
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1198 30 μL
50-167-3955 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1198 Supplier OriGene Supplier No. TA502961S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EB2 (MAPRE2) Monoclonal antibody specifically detects EB2 (MAPRE2) in Canine, Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen EB2 (MAPRE2)
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5F3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MAPRE2
Gene Accession No. Q15555, Q3B8Q0
Gene Alias AI314113; APC-binding protein EB1; APC-binding protein EB2; C820009F03Rik; CSCSC2; D18Abb1e; EB1; EB2; End-binding protein 2; Mapre2; microtubule associated protein RP/EB family member 2; microtubule-associated protein RP/EB family member 2; microtubule-associated protein, RP/EB family, member 2; RP1; T-cell activation protein, EB1 family; T-cell activation protein, EB1 family, member 2
Host Species Mouse
Immunogen Full length human recombinant protein of human MAPRE2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10982, 490487, 679221
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.