Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EB2 (MAPRE2) Monoclonal Antibody (OTI7C10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502871

Description
The protein encoded by this gene shares significant homology to the adenomatous polyposis coli (APC) protein-binding EB1 gene family. The function of this protein is unknown; however, its homology suggests involvement in tumorigenesis of colorectal cancers and proliferative control of normal cells. This gene may belong to the intermediate/early gene family, involved in the signal transduction cascade downstream of the TCR. Alternative splicing results in multiple transcript variants.Specifications
MAPRE2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q15555 | |
MAPRE2 | |
Full length human recombit protein of human MAPRE2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI7C10 | |
Unconjugated | |
MAPRE2 | |
CSCSC2, EB1, EB2, RP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10982 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction