Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAPRE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545912
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545912 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545912 Supplier Invitrogen™ Supplier No. PA545912
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LLCQEHGQEN DDLVQRLMDI LYASEEHEGH TEEPEAEEQA HEQQPPQQEE Sequence homology: Cow: 86%; Dog: 79%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 86%; Yeast: 75%.

The protein encoded by this gene shares significant homology to the adenomatous polyposis coli (APC) protein-binding EB1 gene family. The function of this protein is unknown; however, its homology suggests involvement in tumorigenesis of colorectal cancers and proliferative control of normal cells. This gene may belong to the intermediate/early gene family, involved in the signal transduction cascade downstream of the TCR. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAPRE2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MAPRE2
Gene Accession No. Q15555
Gene Alias AI314113; APC-binding protein EB1; APC-binding protein EB2; C820009F03Rik; CSCSC2; D18Abb1e; EB1; EB2; End-binding protein 2; Mapre2; microtubule associated protein RP/EB family member 2; microtubule-associated protein RP/EB family member 2; microtubule-associated protein, RP/EB family, member 2; RP1; T-cell activation protein, EB1 family; T-cell activation protein, EB1 family, member 2
Gene Symbols MAPRE2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human MAPRE2 (aa 277-326).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10982
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.