Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAPRE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109940
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109940 Supplier Invitrogen™ Supplier No. PA5109940
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAVNVYSTSI TQETMSRHDI IAWVNDIVSL NYTKVEQLCS GAAYCQFMDM LFPGCISLKK VKFQAKLEHE YIHNFKLLQA SFKRMNVDKV IPVEKLVKGR FQDNLDFIQW FKKFYDANYD GKEYDPVEAR QGQDAIPPPD PGEQIFNLPK KSHHANSPTA GAAKSSPAAK PGSTPSRPSS AKRASSSGSA SKSDKDLETQ VIQLNEQVHS LKLALEGVEK ERDFYFGKLR EIELLCQEHG QENDDLVQRL MDILYASEEH EGHTEEPEAE EQAHEQQPPQ QEEY.

The protein encoded by this gene shares significant homology to the adenomatous polyposis coli (APC) protein-binding EB1 gene family. The function of this protein is unknown; however, its homology suggests involvement in tumorigenesis of colorectal cancers and proliferative control of normal cells. This gene may belong to the intermediate/early gene family, involved in the signal transduction cascade downstream of the TCR. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAPRE2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MAPRE2
Gene Accession No. Q15555, Q3B8Q0, Q8R001
Gene Alias AI314113; APC-binding protein EB1; APC-binding protein EB2; C820009F03Rik; CSCSC2; D18Abb1e; EB1; EB2; End-binding protein 2; Mapre2; microtubule associated protein RP/EB family member 2; microtubule-associated protein RP/EB family member 2; microtubule-associated protein, RP/EB family, member 2; RP1; T-cell activation protein, EB1 family; T-cell activation protein, EB1 family, member 2
Gene Symbols MAPRE2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-327 of human MAPRE2 (NP_001137298.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10982, 212307, 679221
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.