Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EB3 Antibody (KT36), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP323294
Description
EB3 Monoclonal antibody specifically detects EB3 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
EB3 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
KT36 | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
EB1 protein family member 3, EB3EBF3-S, End-binding protein 3, microtubule-associated protein RP/EB family member 3, microtubule-associated protein, RP/EB family, member 3, RP3EBF3APC binding protein | |
Clone KT36 was generated from a GST fusion protein containing human EB3. This sequence is highly conserved in rat and mouse EB3. [UniProt# Q9UPY8] | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, GPCR, Signal Transduction | |
22924 | |
Store at 4°C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction