Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EB3, p Ser176 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323300
Description
EB3 Polyclonal antibody specifically detects EB3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
EB3 | |
Polyclonal | |
Western Blot 1:500 | |
EB1 protein family member 3, EB3EBF3-S, End-binding protein 3, microtubule-associated protein RP/EB family member 3, microtubule-associated protein, RP/EB family, member 3, RP3EBF3APC binding protein | |
Phospho-EB3 (Ser-176) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding serine 176 in human EB3. This peptide sequence is highly conserved in mouse and rat EB3. This site is conserved in EB2 but the neighboring amino acids are not well conserved. [UniProt# Q9UPY8] | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, GPCR, Signal Transduction | |
22924 | |
Store at -20°C. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction