Learn More
Invitrogen™ EBAG9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579183
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Testis Tissue, 22RV1 whole cell, HELA whole cell, MCF-7 whole cell, JURKAT whole cell. IHC: mouse testis tissue, rat testis tissue, human prostatic cancer tissue.
This gene was identified as an estrogen-responsive gene. Regulation of transcription by estrogen is mediated by estrogen receptor which binds to the estrogen-responsive element found in the 5'-flanking region of this gene. The encoded protein is a tumor-associated antigen that is expressed at high frequency in a variety of cancers. Two transcript variants differing in the 5' UTR, but encoding the same protein, have been identified for this gene.
Specifications
EBAG9 | |
Polyclonal | |
Unconjugated | |
EBAG9 | |
AI835379; BAG9; cancer associated surface antigen; cancer associated surface antigen RCAS1; cancer-associated surface antigen RCAS1; EB9; EBAG9; estrogen receptor binding fragment associated gene 9; estrogen receptor binding site associated, antigen, 9; estrogen receptor-binding fragment-associated gene 9; estrogen receptor-binding fragment-associated gene 9 protein; HGNC:3123; PDAF; Rcas1; receptor binding cancer antigen expressed on SiSo cells; Receptor-binding cancer antigen expressed on SiSo cells | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
299864, 55960, 9166 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
O00559, Q5PQP2, Q9D0V7 | |
EBAG9 | |
E.coli-derived human EBAG9 recombinant protein (Position: R31-S213). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.