Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBAG9/RCAS1 Antibody (CPTC-EBAG9-1), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308859AF350
Description
EBAG9/RCAS1 Monoclonal antibody specifically detects EBAG9/RCAS1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
EBAG9/RCAS1 | |
Monoclonal | |
Alexa Fluor 350 | |
Cancer-associated surface antigen RCAS1, EB9cancer associated surface antigen, estrogen receptor binding site associated, antigen, 9, Estrogen receptor-binding fragment-associated gene 9 protein, RCAS1PDAF, receptor-binding cancer antigen expressed on SiSo cells | |
Recombinant human full-length EBAG9/RCAS1 protein (Uniprot: O00559) | |
100 μg | |
Apoptosis | |
9166 | |
Store at 4°C in the dark. | |
IgG2c κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
CPTC-EBAG9-1 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction