Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBAG9/RCAS1 Rabbit anti-Human, Clone: EBAG9/7033R, Novus Biologicals™

Rabbit Monoclonal Antibody
$274.50 - $527.00
Specifications
Antigen | EBAG9/RCAS1 |
---|---|
Clone | EBAG9/7033R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
EBAG9/RCAS1 Monoclonal antibody specifically detects EBAG9/RCAS1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
EBAG9/RCAS1 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis | |
10 mM PBS with 0.05% BSA | |
Cancer-associated surface antigen RCAS1, EB9cancer associated surface antigen, estrogen receptor binding site associated, antigen, 9, Estrogen receptor-binding fragment-associated gene 9 protein, RCAS1PDAF, receptor-binding cancer antigen expressed on SiSo cells | |
Recombinant human full-length EBAG9/RCAS1 protein (Uniprot: O00559) | |
Primary | |
Store at 4°C. |
EBAG9/7033R | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
O00559 | |
9166 | |
IgG | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title