Learn More
Ebastine
Description
Ebastine is a second-generation H1 receptor antagonist that is indicated mainly for allergic rhinitis and chronic idiopathic urticaria. It is available in 10 and 20 mg tablets and as fast-dissolving tablets, as well as in pediatric syrup. It has a recommended flexible daily dose of 10 or 20 mg, depending on disease severity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Slightly soluble in chloroform, methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
Specifications
| CAS | 90729-43-4 |
| Melting Point | 84°C to 87°C |
| Color | White |
| Molecular Formula | C32H39NO2 |
| Quantity | 1 g |
| MDL Number | MFCD00865661 |
| Merck Index | 14,3484 |
| Synonym | ebastine, kestine, ebastel, ebastin, kestin, evastel, bactil, ebastinum latin, las w-090, ebastina spanish |
| Solubility Information | Slightly soluble in chloroform,methanol. |
| InChI Key | MJJALKDDGIKVBE-UHFFFAOYSA-N |
| Show More |
Safety and Handling
| RTECSNumber | EL8140000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.