Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBF-1 Antibody (SR2090), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322035100UL
Description
EBF-1 Monoclonal antibody specifically detects EBF-1 in Human, Mouse samples. It is validated for Western BlotSpecifications
EBF-1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
1879 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot | |
SR2090 | |
Western Blot 1:500-1:1000 | |
COE1, Collier, Olf and EBF transcription factor 1, Early B-cell factor, early B-cell factor 1, EBFFLJ41763, O/E-1, OE-1, OLF1FLJ39389, olfactory neuronal transcription factor 1, transcription factor COE1 | |
A synthesized peptide derived from human EBF-1 (Uniprot #: Q9UH73) | |
100 μg | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction