Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBF-1 Antibody (SR2090), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32203550UL
Description
EBF-1 Monoclonal antibody specifically detects EBF-1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| EBF-1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1879 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| SR2090 | |
| Western Blot 1:500-1:1000 | |
| COE1, Collier, Olf and EBF transcription factor 1, Early B-cell factor, early B-cell factor 1, EBFFLJ41763, O/E-1, OE-1, OLF1FLJ39389, olfactory neuronal transcription factor 1, transcription factor COE1 | |
| A synthesized peptide derived from human EBF-1 (Uniprot #: Q9UH73) | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction