Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EBF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529545
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529545 Supplier Invitrogen™ Supplier No. PA529545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: THP-1. Predicted reactivity: Mouse (100%), Rat (96%), Chicken (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Transcriptional activator which recognizes variations of the palindromic sequence 5'-ATTCCCNNGGGAATT-3'. Target protein is a transcription factor that may regulate several olfactory-specific genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBF1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene EBF1
Gene Accession No. Q9UH73
Gene Alias COE1; Collier; Collier, Olf and EBF transcription factor 1; early B cell factor 1; early B-cell factor; early B-cell factor (olfactory neuronal transcription factor 1); early B-cell factor 1; Ebf; EBF transcription factor 1; EBF1; O/E-1; OE-1; OLF1; Olf-1; Olfactory neuronal transcription factor; olfactory neuronal transcription factor 1; transcription factor COE1
Gene Symbols EBF1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human EBF1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1879
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.