Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EBF2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5122021
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122021 20 μL
PIPA5122020 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122021 Supplier Invitrogen™ Supplier No. PA5122021
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Early B-cell factor 2 (EBF2) is a gene located on chromosome 8p11.23, encoding a transcription factor involved in the regulation of gene expression during neural and adipose tissue development. EBF2 belongs to the COE family of transcription factors and plays a pivotal role in cellular differentiation processes. In the nervous system, EBF2 is essential for the development and function of neurons, particularly in regions like the olfactory system. Additionally, EBF2 is involved in adipogenesis, where it regulates genes responsible for the differentiation of brown adipocytes, contributing to thermogenesis and energy metabolism. Dysfunction or altered expression of EBF2 can impact neurological function and metabolic homeostasis, potentially influencing conditions such as obesity, diabetes, and neurodevelopmental disorders. Research into EBF2 is exploring its role in these physiological contexts and the implications of its regulatory mechanisms, offering insights into potential therapeutic approaches.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBF2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene EBF2
Gene Accession No. Q9HAK2
Gene Alias COE2; Collier, Olf and EBF 2; D14Ggc1e; early B cell factor 2; early B-cell factor 2; Ebf2; EBF-2; metencephalon-mesencephalnon-olfactory transcription factor 1; metencephalon-mesencephalon-olfactory transcription factor 1; MET-mesencephalon-olfactory TF1; MET-mesencephalon-olfactory transcription factor 1; Mmot1; O/E-3; OE-3; Olf-1/EBF-like 3; transcription factor COE2
Gene Symbols EBF2
Host Species Rabbit
Immunogen Recombinant EBF2 (Val285-Val536) expressed in E.coli.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64641
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.