Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EBF4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5113949

Catalog No. PIPA5113949


Only null left
Add to cart

Description

Description

Immunogen sequence: GVPGSPSFLN GSTATSPFAK ERLRPRAAPP KLPTPGLPQS PRRGASRPVF For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%; Mouse: 79%.

EBF4 belongs to the COE family and contains 1 IPT/TIG domain. EBF4 is a transcriptional factor which recognizes variations of the palindromic sequence 5'-ATTCCCNNGGGAATT-3'. EBF4 belongs to the conserved Olf/EBF family of helix-loop-helix transcription factors, members of which play important roles in neural development and B-cell maturation (Wang et al., 2002 [PubMed 12139918]).[supplied by OMIM].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EBF4
Polyclonal
Unconjugated
EBF4
COE4; early B cell factor 4; early B-cell factor 3; early B-cell factor 4; early B-cell factor 4-like; EBF family member 4; Ebf3; EBF4; EBF-4; KIAA1442; O/E-4; OE-4; Olf-1; Olf-1/EBF-like 4; RP5-860F19.3; transcription factor COE4
Rabbit
Affinity chromatography
RUO
57593
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9BQW3
EBF4
Synthetic peptide directed towards the C terminal region of human EBF4.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.