Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBF4 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA540626
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540626 Supplier Thermo Scientific Supplier No. PA540626

Rabbit Polyclonal Antibody

Peptide sequence: LYPYIVEFEE EAKNPGLETH RKRKRSGNSD SIERDAAQEA EAGTGLEPGS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%

EBF4 Polyclonal antibody specifically detects EBF4 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBF4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EBF4
Gene Accession No. Q9BQW3
Gene Alias COE4; early B cell factor 4; early B-cell factor 3; early B-cell factor 4; early B-cell factor 4-like; EBF family member 4; Ebf3; EBF4; EBF-4; KIAA1442; O/E-4; OE-4; Olf-1; Olf-1/EBF-like 4; RP5-860F19.3; transcription factor COE4
Gene Symbols EBF4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human EBF4
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57593
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.