Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ebf4 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30395320UL
Description
Ebf4 Polyclonal specifically detects Ebf4 in Human, Mouse samples. It is validated for Western Blot.Specifications
Ebf4 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
57593 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
early B-cell factor 4, EBF-4, olf-1/EBF-like 4, transcription factor COE4 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human EBF4 (NP_001103984.1). | |
20 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction