Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBI3 Antibody (15k8D10) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 11 publications

$243.50 - $487.50

Specifications

Antigen IL-27/IL-35 EBI3 Subunit
Clone 15k8D10
Concentration 1.0 mg/mL
Dilution Western Blot 3-5ug/ml∼, Immunohistochemistry reported in scientific literature (PMID 27681506), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31601798), Immunohistochemistry-Paraffin 5ug/ml, Tissue Culture Substratum
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227362SS
SDP
View Documents
Novus Biologicals
NBP227362SS
0.025 mg
Each for $243.50
Only null left
Add to Cart
 
NBP227362
SDP
View Documents
Novus Biologicals
NBP227362
0.1 mg
Each for $487.50
Only null left
Add to Cart
 
Description

Description

EBI3 Monoclonal specifically detects EBI3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Tissue Culture Substratum.
Specifications

Specifications

IL-27/IL-35 EBI3 Subunit
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
Unconjugated
Mouse
Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction
NP_000873.2
10148
A full-length human Interleukin-35-Fc fusion protein was used as immunogen for this antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
15k8D10
Western Blot 3-5ug/ml∼, Immunohistochemistry reported in scientific literature (PMID 27681506), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31601798), Immunohistochemistry-Paraffin 5ug/ml, Tissue Culture Substratum
Monoclonal
Purified
RUO
Human
cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta
EBI3
IgG1 κ
Protein G purified
The human Interleukin-35 protein is a dimeric protein consisting of the human IL-12a/p35 subunit (NP_000873.2) and the human EBI3 (NP_005746) protein. Clone 15K8D10 tests as being specific to the EBI3 subunit of the target protein.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.