Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBI3 Antibody (42N1G4), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227356AF488
Description
EBI3 Monoclonal specifically detects EBI3 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
IL-27/IL-35 EBI3 Subunit | |
Monoclonal | |
Alexa Fluor 488 | |
Q14213 | |
EBI3 | |
Full-length recombinant human EBI3 protein full length recombinant human EBI3 (cat # 100-162) | |
0.1 mL | |
Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
10148 | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, Flow Cytometry | |
42N1G4 | |
Western Blot, Flow Cytometry | |
cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction