Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBI3 Antibody (42N1G4), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | IL-27/IL-35 EBI3 Subunit |
|---|---|
| Clone | 42N1G4 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2-5ug/ml∼, Flow Cytometry 0.1-1ug/10^6cells |
| Applications | Western Blot, Flow Cytometry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227356SS
![]() |
Novus Biologicals
NBP227356SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227356
![]() |
Novus Biologicals
NBP227356 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
EBI3 Monoclonal specifically detects EBI3 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
| IL-27/IL-35 EBI3 Subunit | |
| 0.5 mg/mL | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
| Q14213 | |
| 10148 | |
| Full-length recombinant human EBI3 protein full length recombinant human EBI3 (cat # 100-162) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 42N1G4 | |
| Western Blot 2-5ug/ml∼, Flow Cytometry 0.1-1ug/10^6cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta | |
| EBI3 | |
| IgG2b κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title