Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBI3 Antibody (5P10D3), DyLight 650, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP203943C
Description
EBI3 Monoclonal specifically detects EBI3 in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
IL-27/IL-35 EBI3 Subunit | |
Monoclonal | |
DyLight 650 | |
cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry | |
5P10D3 | |
Western Blot, Flow Cytometry | |
EBI3 | |
Full-length recombinant mouse EBI3 protein | |
0.1 mL | |
Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
10148 | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction