Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBI3 Monoclonal Antibody (V1.4C4.22), Rockland™
SDP

Catalog No. 501991456
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1456 25 μL
50-199-1455 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1456 Supplier Rockland Immunochemicals Supplier No. 210301B66S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for mouse EBI3 protein. A BLAST analysis was used to suggest cross-reactivity with EBI3 from mouse sources based on 100% homology with the immunizing sequence. Cross-reactivity with EBI-3 from other sources has not been determined.

IL-27 (Interleukin 27) is a cytokine belonging to the IL-12 family, and is a heterodimeric cytokine composed of EBI3 and p28 subunits. IL-27 is produced by activated dendritic cells and macrophages in response to Toll-like receptor ligands and pro-inflammatory cytokines. IL-27 promotes CD4+ T cell differentiation to the Th1 lineage by inducing expression of the transcription factor T-bet and up regulating IL-12R beta2, which in turn suppresses Th2 and Th17 differentiation and proliferation. IL-27 is a recently discovered member of the IL-6/IL-12 family of proinflammatory and immunoregulatory cytokines. IL-27 acts by binding to its receptor WSX-1 and gp130 which results in the activation of a Jak/STAT signaling cascade, suggesting IL-27 involvement in the regulation of immune processes. It has been suggested that IL-27 can also be used as a therapeutic agent against cancer as it can also induce tumor-specific anti-tumor activity mediated through CD8+ T-cells, IFN-gamma, and T-bet. In humans, the gene is located on the p arm of chromosome 16.

Specifications

Antigen IL-35 EBI3
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone V1.4C4.22
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene IL27
Gene Accession No. O35228, Q8K3I6
Gene Alias cytokine receptor; EBI3; EBI-3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; Epstein-Barr virus induced gene 3 (similar to Interleukin-27 beta chain precursor); Epstein-Barr virus-induced gene 3 protein; Epstein-Barr virus-induced gene 3 protein homolog; Il27; IL-27; IL27 p28; IL-27 p28 subunit; IL27 subunit; IL-27 subunit alpha; IL-27 subunit beta; Il27a; IL-27A; IL-27-A; IL27-A; IL27B; IL-27B; IL27p28; IL-27p28; IL30; IL35 subunit; ILN; Interleukin; interleukin 27; interleukin 30; Interleukin-27 subunit alpha; Interleukin-27 subunit beta; Interleukin-30; MGC71873; M-IL-27 (p28); p28; RGD1561420
Gene Symbols Ebi3, IL27
Host Species Mouse
Immunogen Anti-EBI-3 (MOUSE) Monoclonal Antibody was produced in mouse by repeated immunizations with mature full length recombinant mouse EBI-3 produced in E.coli followed by hybridoma development.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246779, 50498
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.