Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ebio1
SDP

Catalog No. 813750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-375-0 50 mg
81-371-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 81-375-0 Supplier Tocris Bioscience™ Supplier No. 8137/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

KCNQ2 activator

Ebio1 is an activator of KCNQ2 voltage-gated potassium channels (EC50 = 247 nM). Ebio1 activates KCNQ2 by increasing both the open probability and the conductance of the channel. Ebio1 has no effect on the voltage sensitivity of KCNQ1, KCNQ3, KCNQ4 and KCNQ5.

Specifications

Chemical Name or Material N-(1,2-Dihydro-5-acenaphthylenyl)-4-fluorobenzamide
CAS 339287-36-4
Quantity 50 mg
Target Voltage-gated Potassium Channel Activators
Molecular Formula C19H14FNO
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.