Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Annexin V Apoptosis Detection Kits

Catalog No. 509280
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
20 Tests
200 Test
200 Tests
300 Tests
50 Tests
Conjugate:
APC, Propidium Iodide
Biotin, Propidium Iodide
FITC, Propidium Iodide
PCP-eFluor 710
PE, 7-AAD
PE/Cy7
eFluor 450, 7-AAD
Excitation/Emission:
405, 546/450, 647
488, 535/520, 617
650, 535/660, 617
NA, 535/NA, 617
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Excitation/Emission
50-928-0 50 Tests PCP-eFluor 710 -
50-927-5 200 Tests FITC, Propidium Iodide 488, 535/520, 617
50-927-6 50 Tests eFluor 450, 7-AAD 405, 546/450, 647
50-927-7 200 Tests eFluor 450, 7-AAD 405, 546/450, 647
50-927-9 200 Tests APC, Propidium Iodide 650, 535/660, 617
50-928-1 200 Test PCP-eFluor 710 -
50-929-7 100 Tests FITC, Propidium Iodide 488, 535/520, 617
50-929-9 20 Tests FITC, Propidium Iodide 488, 535/520, 617
50-930-1 300 Tests FITC, Propidium Iodide 488, 535/520, 617
50-246-343 50 Tests PE, 7-AAD -
50-246-344 200 Tests PE, 7-AAD -
50-246-345 50 Tests PE/Cy7 -
50-246-346 200 Tests PE/Cy7 -
50-929-4 100 Tests Biotin, Propidium Iodide NA, 535/NA, 617
50-929-5 20 Tests Biotin, Propidium Iodide NA, 535/NA, 617
50-929-6 300 Tests Biotin, Propidium Iodide NA, 535/NA, 617
50-112-8958 50 Tests FITC, Propidium Iodide 488, 535/520, 617
50-112-9048 50 Tests APC, Propidium Iodide 650, 535/660, 617
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 50-928-0 Supplier Invitrogen™ Supplier No. 88800872
Add to Cart
Edge
Add to Cart

Includes

  • Cat. No. 88-8008-72:
  • 10X Binding Buffer
  • Annexin V PerCP-eFluor™ 710 : 5μL/test, 50 tests Protect from light.
  • Cat. No. 88-8008-74:
  • 10X Binding Buffer
  • Annexin V PerCP-eFluor™ 710 : 5μL/test, 200 tests Protect from light.

Kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane.

Easily distinguish cells undergoing early and late apoptosis with eBioscience and Annexin V Apoptosis Detection kits for flow cytometry analysis. These kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane. In late-stage apoptosis, as cell integrity is lost, both annexin V and the viability dye traverse and bind PS on the interior of the plasma membrane.

Annexins are a family of calcium-dependent phospholipid-binding proteins that preferentially bind phosphatidylserine (PS). Under normal physiologic conditions, PS is predominantly located in the inner leaflet of the plasma membrane. Upon initiation of apoptosis, PS loses its asymmetric distribution across the phospholipid bilayer and is translocated to the extracellular membrane leaflet marking cells as targets of phagocytosis. Once on the outer surface of the membrane, PS can be detected by fluorescently labeled annexin V in a calcium-dependent manner.

In early-stage apoptosis, the plasma membrane excludes viability dyes such as propidium iodide (PI), 7-AAD, or fixable viability dyes such as eFluor 450. These cells will stain with annexin V but not a viability dye, thus distinguishing cells in early apoptosis. However, in late-stage apoptosis, the cell membrane loses integrity, thereby allowing annexin V to also access PS in the interior of the cell. A viability dye can be used to resolve these late-stage apoptotic and necrotic cells (annexin V, viability dye-positive) from the early-stage apoptotic cells (annexin V positive, viability dye-negative).

Current viability stains available with the eBioscience Annexin V Apoptosis Detection kits include FITC, eFluor 450, allophycocyanin (APC), phycoerythrin (PE), PE-cyanine7, and biotin.

TRUSTED_SUSTAINABILITY

Specifications

Description eBioscience Annexin V Apoptosis Detection Kit PerCP-eFluor 710
Quantity 50 Tests
Product Type Apoptosis Detection Kit
Content And Storage • Annexin V Binding Buffer (10X), 1 bottle (30 mL)
• Annexin V PerCP-eFluor 710, 1 vial (0.25 mL)

Note: Fixable Viability Dye eFluor 450 is not recommended for use with Annexin V Apoptosis Detection Kits. 2°C to 8°C. Avoid temperature fluctuations.
Shipping Condition Wet Ice
Conjugate PCP-eFluor 710
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Can I use eFluor 450-conjugated antibodies with eBioscience Annexin V Apoptosis Detection kits?

Yes, eFluor 450-conjugated antibodies are compatible with eBioscience Annexin V Apoptosis Detection kits.

I trypsinized my adherent cells and labeled with annexin V, and now my flow data is showing a high percentage of apoptotic cells even for control, untreated cells. What is the problem?

Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. Allow the cells to recover for about 30 minutes in optimal cell culture conditions and medium after trypsinizing/scraping so that they can recover their membrane integrity before staining. For lightly adherent cell lines, such as HeLa and NIH 3T3, another option is to use non-enzyme treatments like Gibco Cell Dissociation Buffer (Cat. No. 13151014).

Can I detect annexin V staining in an imaging assay?

Annexin V staining is not typically used in imaging experiments; it is a better reagent for flow cytometry analysis. All cells will stain to some extent, so it can be difficult to distinguish a relatively bright annexin V-stained cell from a dimmer non-apoptotic cell. Caspase activation, detected using our CellEvent Caspase 3/7 or Image-iT LIVE Caspase detection kits, is a better method for detecting apoptosis in an imaging assay.

When should I stain adherent cells with annexin V for flow cytometric analysis? Before or after I trypsinize them?

Trypsinize first and then allow the cells to recover about 30 minutes in optimal cell culture conditions and medium before staining with annexin V conjugates. Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing for annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. For lightly adherent cell lines such as HeLa and NIH 3T3, you could use a less harsh (non-enzymatic) dissociation product like Gibco Cell Dissociation Buffer (Cat. No. 13151014).

Can I fix my cells after annexin V staining?

Yes, this is possible. We have established protocols for annexin V staining combined with intracellular staining of lymphocytes that can be found here. The most important step is to leave some binding buffer in the suspension when fixation is started. Compared to staining of live cells, the intensity of the annexin V signal may be somewhat reduced.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.