Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Annexin V Apoptosis Detection Kits

Catalog No. 509294
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
20 Tests
200 Test
200 Tests
300 Tests
50 Tests
Excitation/Emission:
405, 546/450, 647
488, 535/520, 617
650, 535/660, 617
NA, 535/NA, 617
Conjugate:
APC, Propidium Iodide
Biotin, Propidium Iodide
FITC, Propidium Iodide
PCP-eFluor 710
PE, 7-AAD
PE/Cy7
eFluor 450, 7-AAD
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Excitation/Emission Conjugate
50-929-4 100 Tests NA, 535/NA, 617 Biotin, Propidium Iodide
50-927-5 200 Tests 488, 535/520, 617 FITC, Propidium Iodide
50-927-6 50 Tests 405, 546/450, 647 eFluor 450, 7-AAD
50-927-7 200 Tests 405, 546/450, 647 eFluor 450, 7-AAD
50-927-9 200 Tests 650, 535/660, 617 APC, Propidium Iodide
50-928-0 50 Tests - PCP-eFluor 710
50-928-1 200 Test - PCP-eFluor 710
50-929-7 100 Tests 488, 535/520, 617 FITC, Propidium Iodide
50-929-9 20 Tests 488, 535/520, 617 FITC, Propidium Iodide
50-930-1 300 Tests 488, 535/520, 617 FITC, Propidium Iodide
50-246-343 50 Tests - PE, 7-AAD
50-246-344 200 Tests - PE, 7-AAD
50-246-345 50 Tests - PE/Cy7
50-246-346 200 Tests - PE/Cy7
50-929-5 20 Tests NA, 535/NA, 617 Biotin, Propidium Iodide
50-929-6 300 Tests NA, 535/NA, 617 Biotin, Propidium Iodide
50-112-8958 50 Tests 488, 535/520, 617 FITC, Propidium Iodide
50-112-9048 50 Tests 650, 535/660, 617 APC, Propidium Iodide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 50-929-4 Supplier Invitrogen™ Supplier No. BMS500BT100
Add to Cart
Add to Cart

Includes

  • Annexin V-Biotin protein
  • Binding Buffer (4x)
  • Propidium Iodide (20 ug/mL)

Kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane.

Easily distinguish cells undergoing early and late apoptosis with eBioscience and Annexin V Apoptosis Detection kits for flow cytometry analysis. These kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane. In late-stage apoptosis, as cell integrity is lost, both annexin V and the viability dye traverse and bind PS on the interior of the plasma membrane.

Annexins are a family of calcium-dependent phospholipid-binding proteins that preferentially bind phosphatidylserine (PS). Under normal physiologic conditions, PS is predominantly located in the inner leaflet of the plasma membrane. Upon initiation of apoptosis, PS loses its asymmetric distribution across the phospholipid bilayer and is translocated to the extracellular membrane leaflet marking cells as targets of phagocytosis. Once on the outer surface of the membrane, PS can be detected by fluorescently labeled annexin V in a calcium-dependent manner.

In early-stage apoptosis, the plasma membrane excludes viability dyes such as propidium iodide (PI), 7-AAD, or fixable viability dyes such as eFluor 450. These cells will stain with annexin V but not a viability dye, thus distinguishing cells in early apoptosis. However, in late-stage apoptosis, the cell membrane loses integrity, thereby allowing annexin V to also access PS in the interior of the cell. A viability dye can be used to resolve these late-stage apoptotic and necrotic cells (annexin V, viability dye-positive) from the early-stage apoptotic cells (annexin V positive, viability dye-negative).

Current viability stains available with the eBioscience Annexin V Apoptosis Detection kits include FITC, eFluor 450, allophycocyanin (APC), phycoerythrin (PE), PE-cyanine7, and biotin.

TRUSTED_SUSTAINABILITY

Specifications

Description eBioscience Annexin V-Biotin Apoptosis Detection Kit
Quantity 100 Tests
Product Type Apoptosis Detection Kit
Content And Storage • Binding Buffer (4X), 1 bottle (50 mL)
• rh Annexin V-Biotin, 1 vial (0.5 mL)
• Propidium Iodide (20 μg/mL), 1 vial (1.8 mL). 2°C to 8°C. Avoid temperature fluctuations.
Excitation/Emission NA, 535/NA, 617
Shipping Condition Wet Ice
Conjugate Biotin, Propidium Iodide
For Use With (Equipment) Flow Cytometer
Can I use eFluor 450-conjugated antibodies with eBioscience Annexin V Apoptosis Detection kits?

Yes, eFluor 450-conjugated antibodies are compatible with eBioscience Annexin V Apoptosis Detection kits.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.