Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eBioscience™ Immune Response T Cell Activation Induced Markers Kit

Click to view available options
Quantity:
1 kit
Description
- Kit is comprised of a positive control stimulation reagent, buffers, viability dye, antibodies, and compensation beads
- Use to stimulate and detect expression of CD69, CD134 (OX-40), and CD137 (4-1BB) on human T cells by flow cytometry
- Unique spectral fingerprint is designed to provide the brightest signal for flow cytometry experiments
- Complete kit packaged in single-use vials for ease of use, and to ensure stability performance over time
- Extensive validation and protocol optimization for consistent results
- Stimulation, fixation, permeabilization, and staining are performed in 96-well, round-bottom plates
- Alternatively, cells may be cultured in bulk and transferred to 12 x 75 mm test tubes

Specifications
Specifications
Content And Storage | Antibodies, compensation beads, viability stain, buffers Store at 4°C and –20°C. |
Quantity | 1 kit |
Regulatory Status | RUO |
Shipping Condition | On Ice |
For Use With (Equipment) | Flow Cytometry |
For Use With (Application) | T Cell Activation Induced Marker Analysis and Research |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction