Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Immune Response T Cell Receptor Signaling Kit

Catalog No. A53422
Encompass_Preferred
Click to view available options
Quantity:
1 Kit

Kit is comprised of positive control stimulation reagents, buffers, viability dye, antibodies, and compensation beads needed to stimulate and detect phosphorylation of CD247 (CD3 zeta) on tyrosine 142 (Tyr142) in human T cells by flow cytometry.

  • Kit is comprised of positive control stimulation reagents, buffers, viability dye, antibodies, and compensation beads
  • Use to stimulate and detect phosphorylation of CD247 (CD3 zeta) on tyrosine 142 (Tyr142) in human T cells by flow cytometry
  • Unique spectral fingerprint is designed to provide the brightest signal for flow cytometry experiments
  • Complete kit packaged in single-use vials for ease of use, and to ensure stability performance over time
  • Extensive validation and protocol optimization for consistent results
  • Stimulation, fixation, permeabilization, and staining are performed in 96-well, round-bottom plates
  • Alternatively, cells may be cultured in bulk and transferred to 12 x 75 mm test tubes
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Antibodies, compensation beads, viability stain, buffers
    Store at 4°C and –20°C.
    For Use With (Application) T Cell Receptor Signaling Analysis and Research
    For Use With (Equipment) Flow Cytometry
    Quantity 1 Kit
    Regulatory Status RUO
    Shipping Condition On Ice
    Product Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.