Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBLN2 Antibody (OTI2H1), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271986AF350
Description
EBLN2 Monoclonal specifically detects EBLN2 in Human samples. It is validated for Western Blot.Specifications
EBLN2 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI2H1 | |
Western Blot | |
EBLN2 | |
Full length recombinant protein of human EBLN2 (NP_060499) produced in E.coli. | |
0.1 mL | |
55096.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction