Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EBLN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564385
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564385 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564385 Supplier Invitrogen™ Supplier No. PA564385
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LYAYVNSHSL FVWVCDRSYK RSFRPMILNK IKELSRNQFS TMSHLRKDSQ PSSPGDDAM Highest antigen sequence identity to the following orthologs: Mouse - 25%, Rat - 29%.

May act as an RNA-binding protein. The C-terminal region is highly homologous to the bornavirus nucleocapsid N protein that binds viral RNA and oligomerizes. The viral protein also possesses a nuclear import and a nuclear export signal. These 2 signals seem absent in EBLN-2 supporting an unrelated function in Human.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBLN2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EBLN2
Gene Accession No. Q6P2I7
Gene Alias EBLN-1; EBLN2; EBLN-2; endogenous Borna-like N element-1; endogenous Borna-like N element-2; Endogenous Bornavirus-like nucleoprotein 2; GK006; ikappaBalpha; I-kappa-B-alpha; Ikba; ikB-alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells, alpha; NF-kappa-B inhibitor alpha; Nfkbia; nuclear factor of kappa light chain gene enhancer in B-cells inhibitor, alpha; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha; RL/IF-1
Gene Symbols EBLN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EBLN2. Recombinant protein control fragment (Product #RP-104020).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55096
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.