Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBNA 3A Rabbit anti-Virus, Polyclonal, Bioss
SDP

Catalog No. 501980533
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0533 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0533 Supplier Bioss Supplier No. BS0820R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EBNA3A is a latent viral nuclear protein expressed in Epstein Barr Virus (EBV) transformed lymphoblastic cell lines. It is also found in some immunoblastic lymphomas in vivo. This viral nuclear protein is essential for EBV mediated transformation of B lymphocytes. The EBNA3A functions as a transcriptional regulator though the target genes are currently unknown.

Specifications

Antigen EBNA 3A
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from EBV (Human herpesvirus 4) EBNA 3A .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.