Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBNA1 Antibody (E1-2.5), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500312MFV610
Description
EBNA1 Monoclonal antibody specifically detects EBNA1 in Virus samples. It is validated for ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
EBNA1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
ELISA, Immunocytochemistry | |
E1-2.5 | |
ELISA, Immunocytochemistry/ Immunofluorescence | |
BKRF1, Epstein-Barr nuclear antigen 1, HHV4gp098, nuclear antigen EBNA1 | |
Synthetic peptide corresponding to the repetitive Gly-Ala region of EBNA-1. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction